missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Oxytetracycline ReadyMade™ Solution

Product Code. 30044021
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044021 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044021 Supplier TOKU-E™ Supplier No. O04910x1ML
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxytetracycline ReadyMade™ Solution is provided as a sterile-filtered solution of Oxytetracycline Hydrochloride in water.

  • CAS Number: 2058-46-0
  • Molecular Formula: C22H24N2O9 · HCl
  • Molecular Weight: 496.89 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Synthetic
  • Spectrum: Oxytetracycline is a broad-spectrum antibiotic which targets Gram-positive and Gram-negative species as well as a few Mycoplasma species
  • Mechanism of Action: The mechanism of Oxytetracycline involves diffusing through a cell and binding to the 30s ribosomal subunit preventing peptide elongation and ultimately inhibiting protein synthesis. Resistance to Oxytetracycline can be a result of inactivation by cell enzymes or pumping the antibiotic out of the cell upon entering

Specifications

CAS 2058-46-0
Chemical Name or Material Oxytetracycline HCl
Color Colorless
Content And Storage -20°C
Description Oxytetracycline ReadyMade™ Solution 1 mg/mL in Water
Concentration 1.0 to 1.05 mg/mL
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Media Supplement, Plant Biology Application
Product Line ReadyMade™ Solutions
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.