missing translation for 'onlineSavingsMsg'
Learn More

p-Aminobenzoic Acid, 99+%, MP Biomedicals

Product Code. 11415052 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11415052 1 g 1g
11425052 5 g 5g
11435052 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11415052

Brand: MP Biomedicals 0219461901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 137.1
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material p-Aminobenzoic Acid, 99+%, Free Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.