missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine, 99%

Product Code. p-61020 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10446900

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-94-9 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-94-9, 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 56.0°C to 59.0°C
Color Brown to Gray
Density 1.0600g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Quantity 5 g
Beilstein 13, 435
Merck Index 14, 667
Specific Gravity 1.06
Solubility Information Solubility in water: 21g/L (20°C). Other solubilities: freely soluble in methanol, ethanol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99%
Physical Form Crystalline Powder, Granules or Flakes
Chemical Name or Material p-Anisidine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Anisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : p-Anisidine, o-Anisidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.