missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine hydrochloride, 99%

Product Code. 10697931 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10697931

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20265-97-8 | C7H9NO·HCl | 159.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20265-97-8
Molecular Formula C7H9NO·HCl
Molecular Weight (g/mol) 159.61
MDL Number MFCD00036388
InChI Key VQYJLACQFYZHCO-UHFFFAOYSA-N
Synonym p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride
PubChem CID 2734956
IUPAC Name 4-methoxyaniline;hydrochloride
SMILES COC1=CC=C(C=C1)N.Cl

Specifications

Melting Point 216.0°C to 219.0°C
Color Brown-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2C6H4OCH3·HCl
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 159.61
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine hydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Anisidine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.