missing translation for 'onlineSavingsMsg'
Learn More

p-Anisylchlorodiphenylmethane, 97%

Product Code. 10714281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10714281 100 g Glass bottle 100g
10429070 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10714281 Supplier Thermo Scientific Acros Supplier No. 156561000
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14470-28-1 | C20H17ClO | 308.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14470-28-1
Molecular Formula C20H17ClO
Molecular Weight (g/mol) 308.8
MDL Number MFCD00000814
InChI Key OBOHMJWDFPBPKD-UHFFFAOYSA-N
Synonym 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
PubChem CID 84462
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl

Specifications

Melting Point 121.0°C to 125.0°C
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula CH3OC6H4C(C6H5)2Cl
Quantity 100 g
Beilstein 06, 713
Formula Weight 308.8
Percent Purity 97%
Chemical Name or Material p-Anisylchlorodiphenylmethane, 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.