missing translation for 'onlineSavingsMsg'
Learn More

p-Benzoquinone dioxime, 95%

Product Code. 11488157
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488157 250 g 250g
11478157 50 g 50g
11468157 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17532.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-11-3 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-11-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00063636
InChI Key LNHURPJLTHSVMU-CGXWXWIYSA-N
Synonym 1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
PubChem CID 7737
IUPAC Name N-(4-nitrosophenyl)hydroxylamine
SMILES ON=C1C=CC(C=C1)=NO

Specifications

Melting Point ∼223°C (decomposition)
Density 1.49
Quantity 250 g
Beilstein 2043234
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material p-Benzoquinone dioxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.