missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ P-Cadherin Antibody (PF-03732010), Novus Biologicals™
Human Monoclonal Antibody
206.00 € - 920.00 €
Specifications
| Antigen | P-Cadherin |
|---|---|
| Clone | PF-03732010 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045710
|
Novus Biologicals™
NBP3-28548-100ug |
100 μg |
349.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046632
|
Novus Biologicals™
NBP3-28548-1mg |
1 mg |
920.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046257
|
Novus Biologicals™
NBP3-28548-50ug |
50 μg | |||||||
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| P-Cadherin | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cancer | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin | |
| CDH3/ P-cadherin | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| PF-03732010 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P22223 | |
| 1001 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title