missing translation for 'onlineSavingsMsg'
Learn More

p-Chlorophenol, MP Biomedicals

Product Code. 11407504 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407504 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407504 Supplier MP Biomedicals Supplier No. 0520559680
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 106-48-9
Molecular Formula C6H5ClO
Quantity 100 g
Synonym p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
InChI Key WXNZTHHGJRFXKQ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1O)Cl
IUPAC Name 4-chlorophenol
Molecular Weight (g/mol) 128.555
PubChem CID 4684
ChEBI CHEBI:28078
Formula Weight 128.56
Chemical Name or Material p-Chlorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.