missing translation for 'onlineSavingsMsg'
Learn More

p-Coumaraldehyde, TRC

Product Code. 30351018
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351018 250 mg 250mg
30363320 100 mg 100mg
30355810 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351018 Supplier Toronto Research Chemicals Supplier No. TRCC755450250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2538-87-6
Molecular Formula C9 H8 O2
Molecular Weight (g/mol) 148.16
Synonym 3-(4-Hydroxyphenyl)-2-propenal, 4-Hydroxycinnamaldehyde, p-Coumaraldehyde, p-Coumaroyl aldehyde, p-Hydroxycinnamaldehyde
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enal
SMILES Oc1ccc(\C=C\C=O)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H8O2/c10-7-1-2-8-3-5-9(11)6-4-8/h1-7,11H/b2-1+
Formula Weight 148.0524
Chemical Name or Material p-Coumaraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.