missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 0.99%, Honeywell™

Product Code. 15617790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass bottle
Unit Size:
100g
2kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617790 5 g Glass bottle 5g
15687780 100 g Glass bottle 100g
15607790 500 g Glass bottle 500g
15697780 2 kg Glass bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15617790 Supplier Honeywell Research Chemicals Supplier No. C857515G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O

Specifications

Melting Point 32°C to 35°C
Color Colorless
Density 1.034g/mL at 25°C
Boiling Point 202°C
Flash Point 85°C (185°F)
Ignition Point 1038°F
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 5 g
UN Number UN3455
Beilstein 1305151
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity 0.99
Chemical Name or Material p-Cresol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.