missing translation for 'onlineSavingsMsg'
Learn More

p-(Dimethylamino)benzaldehyde, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15518784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15518784 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518784 Supplier Avantor J.T.Baker Supplier No. J41805
  • ONLINE EXCLUSIVE PRICE
    260.00 €
    226.45 € / 100g
    Save 33.55 €
    12% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Packaging Poly Bottle
Quantity 100 g
Formula Weight 149.19
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material p-(Dimethylamino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.