Learn More
Invitrogen™ P-Glycoprotein Recombinant Rabbit Monoclonal Antibody (SN06-42)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532282
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
| P-Glycoprotein | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P06795, P08183, P21447 | |
| ABCB1, Abcb1a, Abcb1b | |
| Recombinant protein within Human P-glycoprotein 1 aa 373-730. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| SN06-42 | |
| Unconjugated | |
| ABCB1 | |
| ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug resistance protein 1; bovine p-glycoprotein; CD243; CLCS; colchicin sensitivity; doxorubicin resistance; ecotropic viral integration site 32; Evi32; GP170; mdr; Mdr1; MDR1A; Mdr1b; Mdr3; mdr-3; multi-drug resistance 3; multidrug resistance p-glycoprotein; multidrug resistance protein 1; Multidrug resistance protein 1A; multidrug resistance protein 1B; Multidrug resistance protein 2; multidrug resistance protein 3; multidrug resistance protein MDR1; multiple drug resistant 1a; P glycoprotein 1; P glycoprotein 3; P-glycoprotein; P-glycoprotein 1; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein/multidrug resistance 1; Pgp; p-gp; PGP1; PGP2; PGY1; Pgy-1; Pgy1-1; PGY2; Pgy3; Pgy-3; Phospholipid transporter ABCB1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 170913, 18669, 18671, 24646, 5243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.