missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxycinnamic acid, 98%, predominantly trans

Product Code. p-144649 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295250 25 g Plastic bottle 25g
10112730 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295250 Supplier Thermo Scientific Acros Supplier No. 121090250
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501-98-4 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-98-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004399
InChI Key NGSWKAQJJWESNS-ZZXKWVIFSA-N
Synonym p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid
PubChem CID 637542
ChEBI CHEBI:32374
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)O

Specifications

Melting Point 214°C
Color Beige to White or Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula HOC6H4CH=CHCO2H
Quantity 25 g
Beilstein 10, 297
Merck Index 14, 2560
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,practically insoluble in benzene and ligroine
Formula Weight 164.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Hydroxycinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • p-Hydroxycinnamic acid, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.