missing translation for 'onlineSavingsMsg'
Learn More

p-Methoxycinnamic acid, 98%, predominantly trans

Product Code. 10113130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113130 25 g Glass bottle 25g
10408860 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113130 Supplier Thermo Scientific Acros Supplier No. 125840250
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 830-09-1 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-09-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 173°C to 175°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CH=CHCO2H
Quantity 25 g
Beilstein 10, 298
Formula Weight 178.19
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material p-Methoxycinnamic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.