missing translation for 'onlineSavingsMsg'
Learn More

P-Naphtholbenzein, Pure, Indicator Grade, Fisher Chemical™

Product Code. 10598810 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
Unit Size:
25g
Product Code. Quantity Packaging unitSize
10598810 25 g Glass Jar 25g
1 options
This item is not returnable. View return policy

Product Code. 10598810

Brand: Fisher Chemical N/0500/46

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

P-Naphtholbenzein, Pure, Indicator Grade, Fisher Chemical, Quantity: 25g, Packaging: Glass Jar, Brown, Melting Point: 230 deg.C, Molecular Weight: 374.439, CAS: 145-50-6, Formula Weight: 374.44g/mol, InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N

TRUSTED_SUSTAINABILITY

Specifications

CAS 145-50-6
Melting Point 230°C
Color Brown
Packaging Glass Jar
Molecular Formula C27H18O2
MDL Number 78492
Quantity 25 g
Synonym p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene
InChI Key VDDWRTZCUJCDJM-PNHLSOANSA-N
SMILES C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
IUPAC Name (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one
Molecular Weight (g/mol) 374.439
PubChem CID 5941340
Formula Weight 374.44g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Naphtholbenzein
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.