missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenol (Yellow Crystals or Powder/Peptide Synthesis), Fisher BioReagents

Product Code. 10774534
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10774534 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Produktkod. 10774534 Leverantör Fisher BioReagents Leverantörsnummer BP6121
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Nitrophenol is commonly used in peptide synthesis procedures.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Nitrophenol
CAS 100-02-7
Melting Point 112°C
Color Brown
pH 4.4
Boiling Point 279°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Molecular Formula C6H5NO3
MDL Number MFCD00007331
Quantity 1 kg
Merck Index 15, 6708
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitrophenol
Molecular Weight (g/mol) 139.11
PubChem CID 980
ChEBI CHEBI:16836
Formula Weight 139.11
Percent Purity ≥99%
Grade Peptide Synthesis
Physical Form Solid
Chemical Name or Material p-Nitrophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.