missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl 2-Acetamido-2-deoxy-(4-O-2-acetamido-2-deoxy-Beta-D-glucopyranosyl)-Alpha-D-galactopyranoside, TRC

Product Code. 30477624
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477624 1 mg 1mg
30474216 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477624 Supplier Toronto Research Chemicals Supplier No. TRCN4990801MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Nitrophenyl 2-Acetamido-2-deoxy-(4-O-2-acetamido-2-deoxy-beta-D-glucopyranosyl)-alpha-D-galactopyranoside
Analyte or Component Names p-Nitrophenyl 2-Acetamido-2-deoxy-(4-O-2-acetamido-2-deoxy-beta-D-glucopyranosyl)-alpha-D-galactopyranoside
Shipping Condition Room Temperature
Molecular Formula C22H31N3O13
Synonym GlcNAc1-β-4GalNAc-α-PNP, β-p-Nitrophenyl Di-N-acetylchitobioside, p-Nitrophenyl-β-N,N'-diacetylchitobioside, p-Nitrophenyl 2-Acetamido-4-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-2-deoxy-β-D-galactopyranoside
Solution Type Neat
SMILES O[C@H]1[C@H](O)[C@@H](NC(C)=O)[C@H](O[C@@H]2[C@H](O)[C@@H](NC(C)=O)[C@@H](OC3=CC=C([N+]([O-])=O)C=C3)O[C@@H]2CO)O[C@@H]1CO
Industry Type Forensics
Molecular Weight (g/mol) 545.49
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.