missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde, 98%

Product Code. 11420213
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
5g
This item is not returnable. View return policy

Product Code. 11420213

Brand: Thermo Scientific Alfa Aesar A11384.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-87-0 | C8H8O | 120.151 g/mol

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O

Specifications

Melting Point -6°C
Boiling Point 204°C
Flash Point 80°C (176°F)
Quantity 500 g
Beilstein 385772
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material p-Tolualdehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.