missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde, 98%

Product Code. 11410213
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410213 100 g 100g
11414010 5 g 5g
11420213 500 g 500g
11430213 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11410213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11384.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-87-0 | C8H8O | 120.151 g/mol

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O

Specifications

Melting Point -6°C
Density 1.018
Boiling Point 204°C
Flash Point 80°C (176°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 385772
Sensitivity Air Sensitive
Solubility Information It is slightly soluble in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material p-Tolualdehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.