missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid, sodium salt

Product Code. 10412293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412293 500 g Glass bottle 500g
10669384 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412293 Supplier Thermo Scientific Acros Supplier No. 421235000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-84-1
Molecular Formula C7H7NaO3S
Molecular Weight (g/mol) 194.18
MDL Number MFCD00798566,MFCD00064388
InChI Key KVCGISUBCHHTDD-UHFFFAOYSA-M
Synonym sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34
PubChem CID 3720192
IUPAC Name sodium;4-methylbenzenesulfonate
SMILES [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >400°C
Color White
Flash Point >500°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Beilstein 11,97
Fieser 11,535; 12,507
Merck Index 14,9533
Solubility Information Solubility in water: soluble.
Formula Weight 194.19
Percent Purity ≥97.5% (ion exchange; alkalimetry)
Physical Form Powder
Chemical Name or Material p-Toluenesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.