missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic anhydride, 95%

Product Code. 10646394 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646394 100 g Glass bottle 100g
10110762 5 g Glass bottle 5g
10068901 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10646394 Supplier Thermo Scientific Acros Supplier No. 367071000
  • ONLINE EXCLUSIVE PRICE
    426.00 €
    413.00 € / 100g
    Save 13.00 €
    3% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4124-41-8 | C14H14O5S2 | 326.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-41-8
Molecular Formula C14H14O5S2
Molecular Weight (g/mol) 326.39
MDL Number MFCD00008548
InChI Key PDVFSPNIEOYOQL-UHFFFAOYSA-N
Synonym p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
PubChem CID 77773
IUPAC Name (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 124.0°C to 126.0°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Beilstein 11, IV, 376
Formula Weight 326.39
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material p-Toluenesulfonic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluenesulfonic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.