missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic anhydride, 97%

Product Code. 11392675
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392675 25 g 25g
11382675 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392675 Supplier Thermo Scientific Alfa Aesar Supplier No. L00160.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4124-41-8 | C14H14O5S2 | 326.381 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-41-8
Molecular Formula C14H14O5S2
Molecular Weight (g/mol) 326.381
MDL Number MFCD00008548
InChI Key PDVFSPNIEOYOQL-UHFFFAOYSA-N
Synonym p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
PubChem CID 77773
IUPAC Name (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 125°C to 130°C
Quantity 25 g
UN Number UN3261
Beilstein 2223702
Sensitivity Moisture sensitive
Formula Weight 326.38
Percent Purity 97%
Chemical Name or Material p-Toluenesulfonic anhydride
compliance-icons
Product Identifier
  • p-Toluenesulfonic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Toluene-p-sulphonicanhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.