Learn More
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Melting Point | 67.0°C to 69.0°C |
| Color | White |
| Boiling Point | 134.0°C (10.0 mmHg) |
| Flash Point | 128°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min |
| Packaging | Glass bottle |
| Linear Formula | CH3C6H4SO2Cl |
| Quantity | 25 g |
| Beilstein | 11,103 |
| Vedi altri risultati |
Sicurezza e movimentazione
- p-Toluenesulfonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.