missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl cyanide, 95%

Product Code. p-7077580 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15325048

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19158-51-1 | C8H7NO2S | 181.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19158-51-1
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.21
InChI Key JONIMGVUGJVFQD-UHFFFAOYSA-N
Synonym tosyl cyanide, p-toluenesulfonyl cyanide, 4-methylbenzenesulfonyl cyanide, p-toluenesulphonyl cyanide, p-toluene sulfonyl cyanide, p-tolylsulfonyl cyanide, p-toluenesulfonylcyanide, benzenesulfonylcyanide, 4-methyl, benzenesulfonyl cyanide, 4-methyl, 4-methylphenyl sulfonylformonitrile
PubChem CID 87946
IUPAC Name (4-methylphenyl)sulfonylformonitrile
SMILES CC1=CC=C(C=C1)S(=O)(=O)C#N

Specifications

Melting Point 47.0°C to 50.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 181.21
Percent Purity 95%
Chemical Name or Material p-Toluenesulfonyl cyanide
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : p-Toluenesulphonyl cyanide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.