missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl isocyanate, 96%

Product Code. 10033423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10033423 500 mL Glass bottle 500mL
10265520 25 mL Glass bottle 25mL
10072380 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10033423 Supplier Thermo Scientific Acros Supplier No. 183075000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methyl-N-(oxomethylidene)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point -2°C
Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 144°C (10.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO2NCO
Refractive Index 1.5330 to 1.535
Quantity 500 mL
Fieser 07,377
Specific Gravity 1.29
Solubility Information Solubility in water: reacts
Formula Weight 197.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material p-Toluenesulfonyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Toluenesulfonyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.