missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic acid, 98%

Product Code. 10705811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
100g
1kg
250g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10705811 1 kg Plastic bottle 1kg
10366590 100 g Plastic bottle 100g
10521771 250 g Plastic bottle 250g
15109193 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10705811 Supplier Thermo Scientific Acros Supplier No. 139060010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-94-5 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 99-94-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002565
InChI Key LPNBBFKOUUSUDB-UHFFFAOYSA-N
Synonym p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
PubChem CID 7470
ChEBI CHEBI:36635
IUPAC Name 4-methylbenzoic acid
SMILES CC1=CC=C(C=C1)C(O)=O

Spécification

Melting Point 179.0°C to 182.0°C
Color Cream-Yellow to White
Boiling Point 274.0°C to 275.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula CH3C6H4CO2H
Quantity 1 kg
Beilstein 09,483
Merck Index 14,9535
Solubility Information Solubility in water: 11.6g/L (98°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Toluic acid, 98%
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.