missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic hydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11464334
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464334 25 g 25g
11474334 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464334 Supplier Thermo Scientific Alfa Aesar Supplier No. A12604.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3619-22-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007607
InChI Key MFFVZXOPRXMVET-UHFFFAOYSA-N
Synonym p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
PubChem CID 77174
IUPAC Name 4-methylbenzohydrazide
SMILES CC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 115°C to 118°C
Quantity 25 g
Beilstein 607602
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material p-Toluic hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.