missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenediamine, 97%

Product Code. 11367925
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367925 50 g 50g
11347925 2 g 2g
11357925 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367925 Supplier Thermo Scientific Alfa Aesar Supplier No. L02995.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 539-48-0 | C8H12N2 | 136.198 g/mol

p-Xylylenediamine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenediamine is

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-48-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00009821
InChI Key ISKQADXMHQSTHK-UHFFFAOYSA-N
Synonym p-xylylenediamine, 1,4-phenylenedimethanamine, 1,4-bis aminomethyl benzene, 1,4-benzenedimethanamine, 1,4-xylylenediamine, p-xylylendiamine, 4-aminomethylbenzylamine, p-xylylendiamine czech, p-phenylenebis methylamine, 4-aminomethyl phenyl methanamine
PubChem CID 68315
IUPAC Name [4-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)CN

Specifications

Melting Point 59°C to 63°C
Boiling Point 230°C (10 mmHg)
Flash Point 134°C (273°F)
Quantity 50 g
UN Number UN3259
Beilstein 2206190
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material p-Xylylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.