missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VDP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530281
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, mouse liver. Predicted reactivity: Human (99%), Mouse (94%), Rat (93%), Chicken (88%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner.
Specifications
| VDP | |
| Polyclonal | |
| Unconjugated | |
| USO1 | |
| 115kDa; General vesicular transport factor p115; P115; protein USO1 homolog; TAP; transcytosis associated protein; transcytosis associated protein p115; transcytosis-associated protein; USO1; USO1 homolog, vesicle docking protein; USO1 vesicle docking factor; USO1 vesicle docking protein homolog; USO1 vesicle transport factor; Vdp; VDP p115; vesicle docking protein p115; vesicle docking protein, 115 kDa; vesicle-docking protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56041, 8615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60763, Q9Z1Z0 | |
| USO1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human VDP/p115. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction