Learn More
IFN-g Rat anti-Mouse, Alexa Fluor 647, Clone: XMG1.2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557735
Description
The XMG1.2 antibody reacts with mouse interferon-γ (IFN-γ) protein. IFN-γ is a pleiotropic cytokine, of approximately 15-17kDa, involved in the regulation of the immune response. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN-γ production is associated with the Th-1 differentiation. The immunogen was recombinant mouse IFN-γ. This is a neutralizing antibody. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: XMG1.2
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse IFN-<special id='307'/>
Flow Cytometry, Intracellular Staining
Specifications
| IFN-g | |
| Monoclonal | |
| 0.2mg/mL | |
| IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| XMG1.2 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Mouse IFN-γ Recombinant Protein | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.