missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99471-50ul
This item is not returnable.
View return policy
Description
P11 Polyclonal antibody specifically detects P11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| P11 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 3.1, endonuclease, polyU-specific, MGC133268, P11,26 serine protease, Placental protein 11, poly(U)-specific endoribonuclease, PP1122 serine protease, Protein endoU, PRSS26, Uridylate-specific endoribonuclease | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human P11. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8909 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction