missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120-catenin Antibody (CTNND1/4383R), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08618AF532
This item is not returnable.
View return policy
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| p120-catenin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
| Recombinant fragment (around aa900-1,000) of human p120-catenin protein (exact sequence is proprietary) (Uniprot: O60716) | |
| 100 μg | |
| Signal Transduction | |
| 1500 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CTNND1/4383R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction