Learn More
p130 Antibody (2C8A8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52551
 View more versions of this product
Product Code. 18200830
  • 552.00 € / 0.10mg
Description
p130 Monoclonal specifically detects p130 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p130
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
FLJ26459, P130, PRB2, RB2, RBR-2, retinoblastoma-like 2 (p130), retinoblastoma-like protein 2,130 kDa retinoblastoma-associated protein, Retinoblastoma-related protein 2
Mouse
128.4 kDa
0.1 mg
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
5934
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, CyTOF
2C8A8
Unconjugated
PBS with 0.05% Sodium Azide
RBL2
Purified recombinant fragment of human RBL2 (AA: 939-1139) expressed in E. Coli.
Protein A or G purified
RUO
Primary
Human
Purified