missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11300-1-AP-20UL
This item is not returnable.
View return policy
Description
P2Y6 belongs to the family of G protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. P2Y6 expression has been documented in various blood cells, bone marrow, brain, colon, heart, kidney, ovary, placenta, spleen,and thymus. ESTs have been isolated from cervix, embryo, eye, kidney, liver/spleen, placenta, and vessel libraries.Specifications
| P2RY6 | |
| Polyclonal | |
| Unconjugated | |
| P2RY6 | |
| P2RY6, P2Y purinoceptor 6, P2Y6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117264, 233571, 5031 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15077, Q63371, Q9ERK9 | |
| P2RY6 | |
| P2RY6 Fusion Protein Ag1820 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur