Learn More
Invitrogen™ P2Y10 Monoclonal Antibody (639329)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA560852
Product Code. 30536123
  • 419.00 € / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
P2Y10 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. Two alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. P2Y10 has been reported to be expressed in lymphoid cells and in monocytes derived from dendritic cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P2Y10
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O00398
P2RY10
NS0 mouse myeloma cell line transfected with human P2RY10.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin)
639329
Unconjugated
P2RY10
5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1
Mouse
Protein A/G
RUO
27334
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized