missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha Antibody (0004), Novus Biologicals™
Rabbit Monoclonal Antibody
428.00 €
Specifications
| Antigen | p38 alpha |
|---|---|
| Clone | 4 |
| Dilution | Western Blot 1:1000-1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616104
|
Novus Biologicals
NBP3-23507-20ul |
20 μg |
428.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691274
|
Novus Biologicals
NBP3-23507-100ul |
100 μg |
428.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p38 alpha Monoclonal antibody specifically detects p38 alpha in Human samples. It is validated for Western BlotSpecifications
| p38 alpha | |
| Western Blot 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A | |
| This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Thr180, Tyr182 of human phospho-p38 alpha. | |
| Protein A purified |
| 4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 1432 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title