missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha Antibody (10B11C8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37312-0.025ml
This item is not returnable.
View return policy
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| p38 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| Mouse | |
| 41.3 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Protein Kinase, Signal Transduction | |
| 1432 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 10B11C8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MAPK14 | |
| Purified recombinant fragment of human MAPK14 (AA: 299-360) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction