missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p41-ARCb Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552119
This item is not returnable.
View return policy
Description
Immunogen sequence: TVCLADADKK MAVATLASET LPLLALTFIT DNSLVAAGHD CFPVLFTYDA AAGMLSFGGR LDVPKQSSQR GLTARERFQN LDKKASSEGG TAAGAGLDSL HKNSVSQISV LSGGKAKCSQ FCTTGMDGGM SIWDVKSLES A Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. This protein also has a role in centrosomal homeostasis by being an activator and substrate of the Aurora A kinase.
Specifications
| p41-ARCb | |
| Polyclonal | |
| Unconjugated | |
| ARPC1B | |
| 41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10095 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15143 | |
| ARPC1B | |
| Recombinant protein corresponding to Human p41-ARCb. Recombinant protein control fragment (Product #RP-102409). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction