Learn More
Epredia™ Lab Vision™ p53 Rabbit Monoclonal Antibody (Clone SP5)
Employ part of a comprehensive antibody portfolio with the Epredia™ p53, Rabbit Monoclonal Antibody (Clone SP5). Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
130.17 € - 671.47 €
Specifications
| Antigen | p53 |
|---|---|
| Clone | SP5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12603697
|
Epredia™
RM-9105-S0 |
100 μL | Concentrated | ||||||
|
12613697
|
Epredia™
RM-9105-S1 |
500 μL | Concentrated |
493.00 € 453.66 € / 500µL Save 39.34 € 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12693687
|
Epredia™
RM-9105-S |
1 mL | Concentrated |
750.00 € 671.47 € / 1mL Save 78.53 € 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12663687
|
Epredia™
RM-9105-R1 |
1 mL | Prediluted | ||||||
|
12683687
|
Epredia™
RM-9105-R7 |
7 mL | Prediluted |
370.00 € 337.39 € / 7mL Save 32.61 € 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12673687
|
Epredia™
RM-9105-RQ |
12 mL | Ready-to-Use | ||||||
Description
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.Host Species: Rabbit
Clone: SP5
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human full-length wild type p53 protein
Molecular Weight: 53kDa
Positive Control: MDA-231 cells, Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
| p53 | |
| Western Blot | |
| Unconjugated | |
| IVD | |
| Recombinant human full-length wild type p53 protein |
| SP5 | |
| Monoclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.