missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p62/SQSTM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-42822SS
This item is not returnable.
View return policy
Description
p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| p62/SQSTM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| 1 mg/mL | |
| Western Blot 1:2000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 35451674), Immunocytochemistry/Immunofluorescence 1:50, Knockout Validated | |
| Q13501 | |
| SQSTM1 | |
| A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 400-450). [Swiss-Prot Q13501] | |
| 0.025 mL | |
| Autophagy, Signal Transduction | |
| 8878 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction