missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ p73 Antibody - C-terminus, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | p73 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067104
|
Novus Biologicals™
NBP3-30843-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066948
|
Novus Biologicals™
NBP3-30843-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p73 Polyclonal antibody specifically detects p73 in Human,Mouse samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| p73 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| Phosphate buffered saline | |
| 7161 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-10 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
| GST-fused recombinant protein derived from the C terminal of the p73 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title