missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p97/VCP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
316.00 € - 500.00 €
Specifications
| Antigen | p97/VCP |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18478310
|
Novus Biologicals
NBP1-81620-25ul |
25 μL |
316.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18427510
|
Novus Biologicals
NBP1-81620 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p97/VCP Polyclonal antibody specifically detects p97/VCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| p97/VCP | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 7415 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LSDDTCSDEKIRMNRVVRNNLRVRLGDVISIQPCPDVKYGKRIHVLPIDDTVEGITGNLFEVYLKPYFLEAYRPIRKGDIFLVRGGMRAVEFKVVETDPSPYCIVAPDTVIHCEGEPIKREDEEESLNEVGYDDIGGCRKQLAQIKEMVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title