missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPN1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93664-0.1ml
This item is not returnable.
View return policy
Description
PABPN1 Polyclonal antibody specifically detects PABPN1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PABPN1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Nuclear poly(A)-binding protein 1, OPMD, PAB2poly(A) binding protein II, PABII, PABP-2, PABP2polyadenylate-binding protein 2, poly(A) binding protein 2, poly(A) binding protein, nuclear 1, Poly(A)-binding protein 2, Poly(A)-binding protein II, poly(A)-binding protein, nuclear 1, Polyadenylate-binding nuclear protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PABPN1 (NP_004634.1). MAAAAAAAAAAGAAGGRGSGPGRRRHLVPGAGGEAGEGAPGGAGDYGNGLESEELEPEELLLEPEPEPEPEEEPPRPRAPPGAPGPGPGSGAPGSQEEEE | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 8106 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction