missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PADI1 Recombinant Protein Antigen

Product Code. 18730804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18730804 5 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18730804 Supplier Novus Biologicals™ Supplier No. NBP237844
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Description Tissue was fixed in formalin immediately after excision, and embedded in paraffin
Quantity 5
Regulatory Status Research use only
Species Human
Specimen Type Tumor
Tissue Condition Malignant Melanoma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.