missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAF1 Antibody (SR2237), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22163-50ul
This item is not returnable.
View return policy
Description
PAF1 Monoclonal antibody specifically detects PAF1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PAF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2237 | |
| Western Blot 1:500-1:2000 | |
| F23149_1, FLJ11123, Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae), Pancreatic differentiation protein 2, PD2hPAF1, RNA polymerase II-associated factor 1 homolog | |
| A synthesized peptide derived from human PAF1 (Uniprot #: Q8N7H5) | |
| 50 μg | |
| Transcription Factors and Regulators | |
| 54623 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction