missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAF1 Recombinant Rabbit Monoclonal Antibody (24GB625)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | PAF1 |
|---|---|
| Clone | 24GB625 |
| Concentration | 0.41 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276331
![]() |
Invitrogen™
MA557147 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275591
|
Invitrogen™
MA557148 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| PAF1 | |
| 0.41 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4V886, Q8K2T8, Q8N7H5 | |
| 361531, 54623, 54624 | |
| A synthesized peptide derived from human PAF1 (aa 1-56). | |
| Antibody |
| 24GB625 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Paf1 | |
| 5730511K23Rik; AA675045; F23149_1; hPAF1; Paf1; PAF1 homolog, Paf1/RNA polymerase II complex component; Paf1, RNA polymerase II associated factor, homolog; Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae); pancreatic differentiation protein 2; PD2; RGD1306219; RNA polymerase II-associated factor 1 homolog | |
| Paf1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title