missing translation for 'onlineSavingsMsg'
Learn More

Pafuramidine, MedChemExpress

Product Code. 16496258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496258 10 mg 10mg
16452387 50 mg 50mg
16416606 100 mg 100mg
16304778 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496258 Supplier Medchem Express Supplier No. HY14932
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pafuramidine (DB289) is an orally bioavailable prodrug of furamidine, which has activity against Pneumocystis pneumonia.

Pafuramidine (DB289) is an orally bioavailable prodrug of furamidine, which has activity against Pneumocystis pneumonia.

Specifications

Chemical Name or Material Pafuramidine
Physical Form Powder
CAS 186953-56-0
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O3
For Use With (Application) COVID-19-immunoregulation
Synonym DB289
Solubility Information DMSO : 33.33 mg/mL (91.47 mM; Need ultrasonic)
SMILES N=C(C1=CC=C(C2=CC=C(C3=CC=C(C(NOC)=N)C=C3)O2)C=C1)NOC
Molecular Weight (g/mol) 364.4
Formula Weight 364.4
Percent Purity 95.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.