missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAICS Mouse anti-Human, Clone: OTI5B6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501469S
This item is not returnable.
View return policy
Description
This gene encodes a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. The gene is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| PAICS | |
| Monoclonal | |
| Unconjugated | |
| PAICS | |
| ADE2, ADE2H1, AIRC, PAIS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5B6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P22234 | |
| PAICS | |
| Full length human recombit protein of human PAICS produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction