missing translation for 'onlineSavingsMsg'
Learn More

Paliperidone, MedChemExpress

Product Code. 16497986
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497986 100 mg 100mg
16421976 500 mg 500mg
16414157 50 mg 50mg
16401356 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497986 Supplier Medchem Express Supplier No. HYA0019
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Paliperidone (9-Hydroxyrisperidone), the major active metabolite of Risperidone, is a dopamine D2 antagonist and 5-HT2A antagonist. Paliperidone is also active as an antagonist at α1 and α2 adrenergic receptors and H1-histaminergic receptors. Paliperidone, a antipsychotic agent, shows efficacy against schizophrenia.

Paliperidone (9-Hydroxyrisperidone), the major active metabolite of Risperidone, is a dopamine D2 antagonist and 5-HT2A antagonist. Paliperidone is also active as an antagonist at α1 and α2 adrenergic receptors and H1-histaminergic receptors. Paliperidone, a antipsychotic agent, shows efficacy against schizophrenia.

Specifications

Chemical Name or Material Paliperidone
CAS 144598-75-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27FN4O3
Quantity 100 mg
Synonym 9-Hydroxyrisperidone
Solubility Information DMSO : 5 mg/mL (11.72 mM; Need ultrasonic)
SMILES CC1=C(C(N2CCCC(C2=N1)O)=O)CCN3CCC(CC3)C4=NOC5=C4C=CC(F)=C5
Molecular Weight (g/mol) 426.48
Formula Weight 426.48
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.