Learn More
Invitrogen™ Palladin Recombinant Rabbit Monoclonal Antibody (24GB4125)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557151
 View more versions of this product
Product Code. 30275821
  • 500.00 € / 100µL
Description
Palladin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin's different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Palladin
Recombinant Monoclonal
Unconjugated
PALLD
2410003B16Rik; 6030492A02; 90kDa actin-associated protein palladin; actin-associated protein palladin; CGI151; CGI-151; FLJ22190; FLJ38193; FLJ39139; involved in cytoskeletal assembly; KIAA0992; MYN; myoneurin; palladin; palladin isoform; palladin pseudogene; palladin, cytoskeletal associated protein; PALLD; PNCA1; Sarcoma antigen NY-SAR-77; SIH002
Rabbit
Affinity Chromatography
RUO
23022, 364558, 72333
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4125
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P0C5E3, Q8WX93, Q9ET54
PALLD
A synthesized peptide derived from human Palladin (aa 1-65).
100 μL
Primary
Human, Mouse, Rat